Tisk…

Anti-PRKCE Mouse Monoclonal Antibody [clone: 2E9]

Dodavatel: OriGene
ORIGCF502438EA 21120 CZK
ORIGCF502438
Anti-PRKCE Mouse Monoclonal Antibody [clone: 2E9]
Protilátky
Recommended Dilutions: Western Blot: 1:1000-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PRKCE
Clonality: Monoclonal
Clone: 2E9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR