Tisk…

Anti-CTH Mouse Monoclonal Antibody [clone: 2D6]

Dodavatel: OriGene
ORIGCF502443EA 21710 CZK
ORIGCF502443
Anti-CTH Mouse Monoclonal Antibody [clone: 2D6]
Protilátky
Recommended Dilutions: Western Blot: 1:200-500; Immunochemistry: 1:150

Type: Primary
Antigen: CTH
Clonality: Monoclonal
Clone: 2D6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR