Tisk…

Anti-HPGD Mouse Monoclonal Antibody [clone: 1E9]

Dodavatel: OriGene
ORIGCF502452EA 21120 CZK
ORIGCF502452
Anti-HPGD Mouse Monoclonal Antibody [clone: 1E9]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: HPGD
Clonality: Monoclonal
Clone: 1E9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR