Tisk…

Anti-PRL Mouse Monoclonal Antibody [clone: 2E4]

Dodavatel: OriGene
ORIGCF500817EA 21710 CZK
ORIGCF500817
Anti-PRL Mouse Monoclonal Antibody [clone: 2E4]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: PRL
Clonality: Monoclonal
Clone: 2E4
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR