Tisk…
Anti-LIN7B Mouse Monoclonal Antibody [clone: 1G1]
Dodavatel: OriGene
ORIGCF501462EA 21120 CZK
ORIGCF501462
Anti-LIN7B Mouse Monoclonal Antibody [clone: 1G1]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: LIN7B
Clonality: Monoclonal
Clone: 1G1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human