Tisk…

Anti-EPM2AIP1 Mouse Monoclonal Antibody [clone: 5G8]

Dodavatel: OriGene
ORIGCF501793EA 21120 CZK
ORIGCF501793
Anti-EPM2AIP1 Mouse Monoclonal Antibody [clone: 5G8]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: EPM2AIP1
Clonality: Monoclonal
Clone: 5G8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR