Tisk…

Anti-CD4 Mouse Monoclonal Antibody [clone: 9G7]

Dodavatel: OriGene
ORIGCF500484EA 21710 CZK
ORIGCF500484
Anti-CD4 Mouse Monoclonal Antibody [clone: 9G7]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:50-100

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 9G7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR