Tisk…

Anti-SCYL3 Mouse Monoclonal Antibody [clone: 2H10]

Dodavatel: OriGene
ORIGCF500492EA 21120 CZK
ORIGCF500492
Anti-SCYL3 Mouse Monoclonal Antibody [clone: 2H10]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50 Flow Cytometry: 1:1000

Type: Primary
Antigen: SCYL3
Clonality: Monoclonal
Clone: 2H10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR