Tisk…

Anti-SLC18A2 Mouse Monoclonal Antibody [clone: 9E11]

Dodavatel: OriGene
ORIGCF500506EA 21120 CZK
ORIGCF500506
Anti-SLC18A2 Mouse Monoclonal Antibody [clone: 9E11]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SLC18A2
Clonality: Monoclonal
Clone: 9E11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR