Tisk…

Anti-HSPA9 Mouse Monoclonal Antibody [clone: 9F8]

Dodavatel: OriGene
ORIGCF500516EA 21120 CZK
ORIGCF500516
Anti-HSPA9 Mouse Monoclonal Antibody [clone: 9F8]
Protilátky
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:50; Flow Cytometry: 1:100

Type: Primary
Antigen: HSPA9
Clonality: Monoclonal
Clone: 9F8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR