Tisk…

Anti-SIRT5 Mouse Monoclonal Antibody [clone: 1G5]

Dodavatel: OriGene
ORIGCF503280EA 21710 CZK
ORIGCF503280
Anti-SIRT5 Mouse Monoclonal Antibody [clone: 1G5]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: SIRT5
Clonality: Monoclonal
Clone: 1G5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR