Tisk…

Anti-SIRT5 Mouse Monoclonal Antibody [clone: 1A6]

Dodavatel: OriGene
ORIGCF503281EA 21710 CZK
ORIGCF503281
Anti-SIRT5 Mouse Monoclonal Antibody [clone: 1A6]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100

Type: Primary
Antigen: SIRT5
Clonality: Monoclonal
Clone: 1A6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR