Tisk…

Anti-CA12 Mouse Monoclonal Antibody [clone: 4G3]

Dodavatel: OriGene
ORIGCF505610EA 21120 CZK
ORIGCF505610
Anti-CA12 Mouse Monoclonal Antibody [clone: 4G3]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: CA12
Clonality: Monoclonal
Clone: 4G3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR