Tisk…

Anti-MAP2K3 Mouse Monoclonal Antibody [clone: 1D3]

Dodavatel: OriGene
ORIGCF505617EA 21120 CZK
ORIGCF505617
Anti-MAP2K3 Mouse Monoclonal Antibody [clone: 1D3]
Protilátky
Recommended Dilutions: Western Blot: 1:200-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: MAP2K3
Clonality: Monoclonal
Clone: 1D3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR