Tisk…

Anti-ZFYVE21 Mouse Monoclonal Antibody [clone: 4A2]

Dodavatel: OriGene
ORIGCF505622EA 21120 CZK
ORIGCF505622
Anti-ZFYVE21 Mouse Monoclonal Antibody [clone: 4A2]
Protilátky
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: ZFYVE21
Clonality: Monoclonal
Clone: 4A2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR