Tisk…

Anti-TBC1D21 Mouse Monoclonal Antibody [clone: 8B7]

Dodavatel: OriGene
ORIGCF503716EA 21710 CZK
ORIGCF503716
Anti-TBC1D21 Mouse Monoclonal Antibody [clone: 8B7]
Protilátky
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: TBC1D21
Clonality: Monoclonal
Clone: 8B7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR