Tisk…

Anti-TMEFF2 Mouse Monoclonal Antibody [clone: 4G2]

Dodavatel: OriGene
ORIGCF504465EA 21120 CZK
ORIGCF504465
Anti-TMEFF2 Mouse Monoclonal Antibody [clone: 4G2]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: TMEFF2
Clonality: Monoclonal
Clone: 4G2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR