Tisk…
Anti-SENP2 Mouse Monoclonal Antibody [clone: 3H10]
Dodavatel: OriGene
ORIGCF504864EA 21120 CZK
ORIGCF504864
Anti-SENP2 Mouse Monoclonal Antibody [clone: 3H10]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000
Type: Primary
Antigen: SENP2
Clonality: Monoclonal
Clone: 3H10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human