Tisk…

Anti-DDT Mouse Monoclonal Antibody [clone: 3A1]

Dodavatel: OriGene
ORIGCF505656EA 21710 CZK
ORIGCF505656
Anti-DDT Mouse Monoclonal Antibody [clone: 3A1]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: DDT
Clonality: Monoclonal
Clone: 3A1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR