Tisk…

Anti-HRAS Mouse Monoclonal Antibody [clone: 1C2]

Dodavatel: OriGene
ORIGCF505667EA 21120 CZK
ORIGCF505667
Anti-HRAS Mouse Monoclonal Antibody [clone: 1C2]
Protilátky
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: HRAS
Clonality: Monoclonal
Clone: 1C2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR