Tisk…

Anti-TBC1D21 Mouse Monoclonal Antibody [clone: 3F1]

Dodavatel: OriGene
ORIGCF503742EA 21710 CZK
ORIGCF503742
Anti-TBC1D21 Mouse Monoclonal Antibody [clone: 3F1]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: TBC1D21
Clonality: Monoclonal
Clone: 3F1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR