Tisk…

Anti-PPAT Mouse Monoclonal Antibody [clone: 1D9]

Dodavatel: OriGene
ORIGCF504508EA 21120 CZK
ORIGCF504508
Anti-PPAT Mouse Monoclonal Antibody [clone: 1D9]
Protilátky
Recommended Dilutions: Western Blot: 1:200-1:1000

Type: Primary
Antigen: PPAT
Clonality: Monoclonal
Clone: 1D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR