Tisk…

Anti-LHX1 Mouse Monoclonal Antibody [clone: 2G3]

Dodavatel: OriGene
ORIGCF504534EA 21120 CZK
ORIGCF504534
Anti-LHX1 Mouse Monoclonal Antibody [clone: 2G3]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: LHX1
Clonality: Monoclonal
Clone: 2G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR