Tisk…

Anti-LRRC25 Mouse Monoclonal Antibody [clone: 1D1]

Dodavatel: OriGene
ORIGCF504917EA 21710 CZK
ORIGCF504917
Anti-LRRC25 Mouse Monoclonal Antibody [clone: 1D1]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: LRRC25
Clonality: Monoclonal
Clone: 1D1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR