Tisk…

Anti-SNAI2 Mouse Monoclonal Antibody [clone: 1H10]

Dodavatel: OriGene
ORIGCF504929EA 21120 CZK
ORIGCF504929
Anti-SNAI2 Mouse Monoclonal Antibody [clone: 1H10]
Protilátky
Recommended Dilutions: Western Blot: 1:200-1:1000

Type: Primary
Antigen: SNAI2
Clonality: Monoclonal
Clone: 1H10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR