Tisk…

Anti-CDC123 Mouse Monoclonal Antibody [clone: 2D4]

Dodavatel: OriGene
ORIGCF505692EA 21120 CZK
ORIGCF505692
Anti-CDC123 Mouse Monoclonal Antibody [clone: 2D4]
Protilátky
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150

Type: Primary
Antigen: CDC123
Clonality: Monoclonal
Clone: 2D4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR