Tisk…

Anti-MDM4 Mouse Monoclonal Antibody [clone: 4B5]

Dodavatel: OriGene
ORIGCF505706EA 21120 CZK
ORIGCF505706
Anti-MDM4 Mouse Monoclonal Antibody [clone: 4B5]
Protilátky
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: MDM4
Clonality: Monoclonal
Clone: 4B5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR