Tisk…

Anti-CDH2 Mouse Monoclonal Antibody [clone: 3B10]

Dodavatel: OriGene
ORIGCF503775EA 21120 CZK
ORIGCF503775
Anti-CDH2 Mouse Monoclonal Antibody [clone: 3B10]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: CDH2
Clonality: Monoclonal
Clone: 3B10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR