Tisk…

Anti-ADIPOQ Mouse Monoclonal Antibody [clone: 1B2]

Dodavatel: OriGene
ORIGCF503801EA 21120 CZK
ORIGCF503801
Anti-ADIPOQ Mouse Monoclonal Antibody [clone: 1B2]
Protilátky
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ADIPOQ
Clonality: Monoclonal
Clone: 1B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR