Tisk…

Anti-HMOX2 Mouse Monoclonal Antibody [clone: 1C2]

Dodavatel: OriGene
ORIGCF503813EA 21710 CZK
ORIGCF503813
Anti-HMOX2 Mouse Monoclonal Antibody [clone: 1C2]
Protilátky
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: HMOX2
Clonality: Monoclonal
Clone: 1C2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR