Tisk…
Anti-MAP2K1 Mouse Monoclonal Antibody [clone: 2A9]
Dodavatel: OriGene
ORIGCF505764EA 21120 CZK
ORIGCF505764
Anti-MAP2K1 Mouse Monoclonal Antibody [clone: 2A9]
Protilátky
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: MAP2K1
Clonality: Monoclonal
Clone: 2A9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Mouse, Rat