Tisk…

Anti-RAB21 Mouse Monoclonal Antibody [clone: 5B12]

Dodavatel: OriGene
ORIGCF505777EA 21710 CZK
ORIGCF505777
Anti-RAB21 Mouse Monoclonal Antibody [clone: 5B12]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: RAB21
Clonality: Monoclonal
Clone: 5B12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR