Tisk…

Anti-MAPK13 Mouse Monoclonal Antibody [clone: 12C6]

Dodavatel: OriGene
ORIGCF505779EA 21120 CZK
ORIGCF505779
Anti-MAPK13 Mouse Monoclonal Antibody [clone: 12C6]
Protilátky
Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100

Type: Primary
Antigen: MAPK13
Clonality: Monoclonal
Clone: 12C6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR