Tisk…

Anti-MAPK13 Mouse Monoclonal Antibody [clone: 2G1]

Dodavatel: OriGene
ORIGCF505852EA 21120 CZK
ORIGCF505852
Anti-MAPK13 Mouse Monoclonal Antibody [clone: 2G1]
Protilátky
Recommended Dilutions: Western Blot: 1:200-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: MAPK13
Clonality: Monoclonal
Clone: 2G1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR