Tisk…

Anti-THUMPD1 Mouse Monoclonal Antibody [clone: 1H6]

Dodavatel: OriGene
ORIGCF505871EA 21710 CZK
ORIGCF505871
Anti-THUMPD1 Mouse Monoclonal Antibody [clone: 1H6]
Protilátky
Recommended Dilutions: Western Blot: 1:200-4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: THUMPD1
Clonality: Monoclonal
Clone: 1H6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR