Tisk…

Anti-TUBAL3 Mouse Monoclonal Antibody [clone: 1D7]

Dodavatel: OriGene
ORIGCF503924EA 21120 CZK
ORIGCF503924
Anti-TUBAL3 Mouse Monoclonal Antibody [clone: 1D7]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: TUBAL3
Clonality: Monoclonal
Clone: 1D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR