Tisk…
Anti-TSC22D1 Mouse Monoclonal Antibody [clone: 4H8]
Dodavatel: OriGene
ORIGCF505062EA 21120 CZK
ORIGCF505062
Anti-TSC22D1 Mouse Monoclonal Antibody [clone: 4H8]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100
Type: Primary
Antigen: TSC22D1
Clonality: Monoclonal
Clone: 4H8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human