Tisk…

Anti-RIOK2 Mouse Monoclonal Antibody [clone: 2F4]

Dodavatel: OriGene
ORIGCF505085EA 21710 CZK
ORIGCF505085
Anti-RIOK2 Mouse Monoclonal Antibody [clone: 2F4]
Protilátky
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: RIOK2
Clonality: Monoclonal
Clone: 2F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR