Tisk…

Anti-CTAG1B Mouse Monoclonal Antibody [clone: 1G9]

Dodavatel: OriGene
ORIGCF505945EA 21120 CZK
ORIGCF505945
Anti-CTAG1B Mouse Monoclonal Antibody [clone: 1G9]
Protilátky
Recommended Dilutions: Western Blot: 1:200-1:1000; Immunofluorescence: 1:100

Type: Primary
Antigen: CTAG1B
Clonality: Monoclonal
Clone: 1G9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR