Tisk…

Anti-CTAG1B Mouse Monoclonal Antibody [clone: 2A10]

Dodavatel: OriGene
ORIGCF505948EA 21120 CZK
ORIGCF505948
Anti-CTAG1B Mouse Monoclonal Antibody [clone: 2A10]
Protilátky
Recommended Dilutions: Western Blot: 1:200-1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: CTAG1B
Clonality: Monoclonal
Clone: 2A10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR