Tisk…

Anti-TRIM38 Mouse Monoclonal Antibody [clone: 2A4]

Dodavatel: OriGene
ORIGCF505964EA 21120 CZK
ORIGCF505964
Anti-TRIM38 Mouse Monoclonal Antibody [clone: 2A4]
Protilátky
Recommended Dilutions: Western Blot: 1:200

Type: Primary
Antigen: TRIM38
Clonality: Monoclonal
Clone: 2A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR