Tisk…

Anti-CTAG1B Mouse Monoclonal Antibody [clone: 9H6]

Dodavatel: OriGene
ORIGCF505974EA 21120 CZK
ORIGCF505974
Anti-CTAG1B Mouse Monoclonal Antibody [clone: 9H6]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: CTAG1B
Clonality: Monoclonal
Clone: 9H6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR