Tisk…

Anti-DPP9 Mouse Monoclonal Antibody [clone: 2D4]

Dodavatel: OriGene
ORIGCF503959EA 21120 CZK
ORIGCF503959
Anti-DPP9 Mouse Monoclonal Antibody [clone: 2D4]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: DPP9
Clonality: Monoclonal
Clone: 2D4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR