Tisk…

Anti-ACSF2 Mouse Monoclonal Antibody [clone: 6E4]

Dodavatel: OriGene
ORIGCF504711EA 21120 CZK
ORIGCF504711
Anti-ACSF2 Mouse Monoclonal Antibody [clone: 6E4]
Protilátky
Recommended Dilutions: Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ACSF2
Clonality: Monoclonal
Clone: 6E4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR