Tisk…

Anti-METT10D Mouse Monoclonal Antibody [clone: 1A2]

Dodavatel: OriGene
ORIGCF504728EA 21710 CZK
ORIGCF504728
Anti-METT10D Mouse Monoclonal Antibody [clone: 1A2]
Protilátky
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: METT10D
Clonality: Monoclonal
Clone: 1A2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR