Tisk…

Anti-TBCC Mouse Monoclonal Antibody [clone: 4C4]

Dodavatel: OriGene
ORIGCF504735EA 21710 CZK
ORIGCF504735
Anti-TBCC Mouse Monoclonal Antibody [clone: 4C4]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: TBCC
Clonality: Monoclonal
Clone: 4C4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR