Tisk…
Anti-CNKSR3 Mouse Monoclonal Antibody [clone: 1D7]
Dodavatel: OriGene
ORIGCF504737EA 21120 CZK
ORIGCF504737
Anti-CNKSR3 Mouse Monoclonal Antibody [clone: 1D7]
Protilátky
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150
Type: Primary
Antigen: CNKSR3
Clonality: Monoclonal
Clone: 1D7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human