Tisk…

Anti-NQO2 Mouse Monoclonal Antibody [clone: 3C11]

Dodavatel: OriGene
ORIGCF504744EA 21120 CZK
ORIGCF504744
Anti-NQO2 Mouse Monoclonal Antibody [clone: 3C11]
Protilátky
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: NQO2
Clonality: Monoclonal
Clone: 3C11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR