Tisk…

Anti-CHN1 Mouse Monoclonal Antibody [clone: 3C6]

Dodavatel: OriGene
ORIGCF505109EA 21120 CZK
ORIGCF505109
Anti-CHN1 Mouse Monoclonal Antibody [clone: 3C6]
Protilátky
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:150

Type: Primary
Antigen: CHN1
Clonality: Monoclonal
Clone: 3C6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR