Tisk…

Anti-MAP2K3 Mouse Monoclonal Antibody [clone: 2G8]

Dodavatel: OriGene
ORIGCF505806EA 21120 CZK
ORIGCF505806
Anti-MAP2K3 Mouse Monoclonal Antibody [clone: 2G8]
Protilátky
Recommended Dilutions: Western Blot: 1:200-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: MAP2K3
Clonality: Monoclonal
Clone: 2G8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR